EMAIL THIS PAGE TO A FRIEND
Journal of Orthopaedic Research : Official Publication of the Orthopaedic Research Society
Mouse synovial mesenchymal stem cells increase in yield with knee inflammation.
PMID 25403845