EMAIL THIS PAGE TO A FRIEND
Canadian journal of microbiology
Zinc biosorption by the purple non-sulfur bacterium Rhodobacter capsulatus.
PMID 25403904
Abstract
This paper presents the first report providing information on the zinc (Zn) biosorption potentialities of the purple non-sulfur bacterium Rhodobacter capsulatus. The effects of various biological, physical, and chemical parameters on Zn biosorption were studied in both the wild-type strain B10 and a strain, RC220, lacking the endogenous plasmid. At an initial Zn concentration of 10 mg·L(-1), the Zn biosorption capacity at pH 7 for bacterial biomass grown in synthetic medium containing lactate as carbon source was 17 and 16 mg Zn·(g dry mass)(-1) for strains B10 and RC220, respectively. Equilibrium was achieved in a contact time of 30-120 min, depending on the initial Zn concentration. Zn sorption by live biomass was modelled, at equilibrium, according to the Redlich-Peterson and Langmuir isotherms, in the range of 1-600 mg Zn·L(-1). The wild-type strain showed a maximal Zn uptake capacity (Qm) of 164 ± 8 mg·(g dry mass)(-1) and an equilibrium constant (Kads) of 0.017 ± 0.00085 L·(mg Zn)(-1), compared with values of 73.9 mg·(g dry mass)(-1) and 0.361 L·mg(-1) for the strain lacking the endogenous plasmid. The Qm value observed for R. capsulatus B10 is one of the highest reported in the literature, suggesting that this strain may be useful for Zn bioremediation. The lower Qm value and higher equilibrium constant observed for strain RC220 suggest that the endogenous plasmid confers an enhanced biosorption capacity in this bacterium, although no genetic determinants for Zn resistance appear to be located on the plasmid, and possible explanations for this are discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
209988
Zinc, dust, <10 μm, ≥98%
Zn
324930
Zinc, powder, <150 μm, 99.995% trace metals basis
Zn
243469
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Zn
31653
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Zn
96454
Zinc, purum, powder
Zn
267619
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Zn
578002
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Zn
356018
Zinc, foil, thickness 0.25 mm, 99.9% trace metals basis
Zn
565148
Zinc, granular, 30-100 mesh, 99%
Zn
267635
Zinc, shot, 5 mm, 99.999% trace metals basis
Zn
267929
Zinc, wire, diam. 1.0 mm, 99.999% trace metals basis
Zn
266361
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Zn
402583
Zinc, shot, <12 mm, 99.99% trace metals basis
Zn
215481
Zinc, mossy, ≥99%
Zn
266353
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Zn
GF86667812
Zinc, foil, 100x100mm, thickness 1.0mm, as rolled, 99.95+%
Zn
716723
Zinc, shot, 99.9999%
Zn
349410
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Zn
215503
Zinc, mossy, 99.99% trace metals basis
Zn
GF60963453
Zinc, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.99+%
Zn
GF63340758
Zinc, foil, 100x100mm, thickness 0.20mm, as rolled, 99.95+%
Zn
749060
Zinc, sputtering target, diam. × thickness 3.00 in. × 0.125 in., 99.995% trace metals basis
Zn
GF59993061
Zinc, foil, light tested, 100x100mm, thickness 0.025mm, as rolled, 99.95+%
Zn
GF37287704
Zinc, wire reel, 100m, diameter 0.25mm, as drawn, 99.99+%
Zn
GF32078853
Zinc, foil, light tested, 150x150mm, thickness 0.05mm, as rolled, 99.95+%
Zn
GF72714102
Zinc, wire reel, 0.1m, diameter 1.0mm, 99.999%
Zn
GF47920305
Zinc, wire reel, 10m, diameter 0.125mm, as drawn, 99%
Zn
GF58911385
Zinc, wire reel, 0.5m, diameter 1.0mm, 99.99+%
Zn
GF99245728
Zinc, foil, 300x300mm, thickness 0.20mm, as rolled, 99.95+%
Zn
GF01301612
Zinc, foil, not light tested, 25x25mm, thickness 0.0025mm, 99.9%
Zn