EMAIL THIS PAGE TO A FRIEND
Lipids
n-3 PUFA esterified to glycerol or as ethyl esters reduce non-fasting plasma triacylglycerol in subjects with hypertriglyceridemia: a randomized trial.
PMID 25403919