EMAIL THIS PAGE TO A FRIEND
Oxidative Medicine and Cellular Longevity
Reduced oxidative stress contributes to the lipid lowering effects of isoquercitrin in free fatty acids induced hepatocytes.
PMID 25404990