EMAIL THIS PAGE TO A FRIEND
Endocrinology
Sex-specific effects of relaxin-3 on food intake and brain expression of corticotropin-releasing factor in rats.
PMID 25406021
Abstract
This study compared the effects of relaxin-3 (RLN3) on food intake, plasma corticosterone, and the expression of corticotropin-releasing factor (CRF) in male and female rats. RLN3 was injected into the lateral ventricle at 25, 200, and 800 pmol concentrations. RLN3 at 25 pmol increased food intake (grams) at 30 and 60 minutes after injection in female but not male rats. Female rats also showed higher increase in relative to body weight (BW) food intake (mg/g BW) for all RLN3 concentrations at 30 minutes and for 800 pmol of RLN3 at 60 minutes. Moreover, RLN3 at 800 pmol significantly increased 24-hour BW gain in female but not male rats. At 60 minutes after administration, 800 pmol of RLN3 produced a significant increase in plasma corticosterone and in the expression of CRF and c-fos mRNAs in the parvocellular paraventricular hypothalamic nucleus (PVN) in male but not female rats. The levels of c-fos mRNA in the magnocellular PVN were increased by RLN3 but did not differ between the sexes. Conversely, expression of CRF mRNA in the medial preoptic area was increased in female rats but was not sensitive to 800 pmol of RLN3. In the bed nucleus of the stria terminalis, 800 pmol of RLN3 significantly increased CRF mRNA expression in female but not male rats. Therefore, female rats showed more sensitivity and stronger food intake increase in response to RLN3. The differential effects of RLN3 on CRF expression in the PVN and bed nucleus of the stria terminalis may contribute to the sex-specific difference in the behavioral response.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
66725
Methylene Blue solution alkaline according to Loeffler, according to Loeffler
C16H18ClN3S