EMAIL THIS PAGE TO A FRIEND
ACS Chemical Biology
Human phosphoglycerate dehydrogenase produces the oncometabolite D-2-hydroxyglutarate.
PMID 25406093
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
236691
(R)-(−)-2-Butanol, 99%
C4H10O