EMAIL THIS PAGE TO A FRIEND
The Journal of antibiotics
Cytotoxic hydroanthraquinones from the mangrove-derived fungus Paradictyoarthrinium diffractum BCC 8704.
PMID 25407145
Abstract
Two new hydroanthraquinones, paradictyoarthrins A (1) and B (2), were isolated from the mangrove-derived fungus Paradictyoarthrinium diffractum BCC 8704. Structures of the new compounds were elucidated by analyses of the NMR spectroscopic and mass spectrometry data. The absolute configuration of 1 was determined by X-ray crystallography. These compounds exhibited cytotoxic activities.