EMAIL THIS PAGE TO A FRIEND
Chemical biology & drug design
Ranking the Binding Energies of p53 Mutant Activators and Their ADMET Properties.
PMID 25407396
Abstract
The guardian of the genome, p53, is the most mutated protein found in all cancer cells. Restoration of wild-type activity to mutant p53 offers promise to eradicate cancer cells using novel pharmacological agents. Several molecules have already been found to activate mutant p53. While the exact mechanism of action of these compounds has not been fully understood, a transiently open pocket has been identified in some mutants. In our study, we docked twelve known activators to p53 into the open pocket to further understand their mechanism of action and rank the best binders. In addition, we predicted the absorption, distribution, metabolism, excretion and toxicity properties of these compounds to assess their pharmaceutical usefulness. Our studies showed that alkylating ligands do not all bind at the same position, probably due to their varying sizes. In addition, we found that non-alkylating ligands are capable of binding at the same pocket and directly interacting with Cys124. The comparison of the different ligands demonstrates that stictic acid has a great potential as a p53 activator in terms of less adverse effects although it has poorer pharmacokinetic properties.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W2020
WR-1065, ≥98% (HPLC)
C5H14N2S · 2HCl