EMAIL THIS PAGE TO A FRIEND
Pharmaceutical Research
Bridging laboratory and large scale production: preparation and in vitro-evaluation of photosensitizer-loaded nanocarrier devices for targeted drug delivery.
PMID 25407544