EMAIL THIS PAGE TO A FRIEND
Environmental monitoring and assessment
Utilization of biochar sorbents for Cd²⁺, Zn²⁺, and Cu²⁺ ions separation from aqueous solutions: comparative study.
PMID 25407990
Abstract
The objective of this study was to study the utilization of two different woody-derived biochars for Cd(2+), Zn(2+), and Cu(2+) ions separation from aqueous solutions. Physicochemical characterization confirmed the main differences in sorbent surface area and cation-exchange capacity. The maximum cadmium, zinc, and copper sorption capacities were 1.99, 0.97, and 2.50 mg g(-1) for biochar (BC) A; 7.80, 2.23, and 3.65 mg g(-1) for BC B. Sorption processes can be affected by time and pH. The most of sorbed cadmium and zinc were bound on exchangeable fractions and copper oxidizable fractions. Chemical modification and FT-IR analyses confirmed the crucial roles of hydroxyl and mainly carboxyl functional groups in sorption processes of Cd(2+), Zn(2+), and Cu(2+) ions by BC A and BC B. The garden wood rests with leaf mass-derived biochar can be utilized as an effective sorbent for bivalent ions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
414891
Cadmium, granular, 30-80 mesh, ≥99%
Cd
265330
Cadmium, granular, ≥99%, 5-20 mesh
Cd
265365
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Cd
202886
Cadmium, shot, 3 mm, 99.999% trace metals basis
Cd
348600
Cadmium, wire, diam. 1.0 mm, 99.999% trace metals basis
Cd
265454
Cadmium, rod, diam. 4.0 mm, 99.999% trace metals basis
Cd
GF61936814
Cadmium, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%
Cd
GF33512688
Cadmium, foil, 200x200mm, thickness 0.5mm, as rolled, 99.99%
Cd
GF64286602
Cadmium, foil, 100x100mm, thickness 1.0mm, as rolled, 99.99%
Cd
GF38539929
Cadmium, foil, not light tested, 100x100mm, thickness 0.025mm, 99.7+%
Cd
GF31881911
Cadmium, rod, 500mm, diameter 5.0mm, as drawn, 99.99+%
Cd
GF72757503
Cadmium, foil, 150x150mm, thickness 0.5mm, as rolled, 99.99%
Cd
GF72955772
Cadmium, rod, 50mm, diameter 40mm, 99.9%
Cd
GF85239096
Cadmium, rod, 1000mm, diameter 5.0mm, as drawn, 99.99+%
Cd
GF30096345
Cadmium, foil, 100x100mm, thickness 2.0mm, as rolled, 99.99%
Cd
GF59435062
Cadmium, foil, 25x25mm, thickness 0.5mm, as rolled, 99.999%
Cd
GF01198762
Cadmium, wire reel, 1m, diameter 0.25mm, hard, 99.99+%
Cd
GF82927064
Cadmium, foil, 50x50mm, thickness 1.0mm, as rolled, 99.999%
Cd
GF32202582
Cadmium, foil, not light tested, 25x25mm, thickness 0.025mm, 99.7+%
Cd
GF86338154
Cadmium, tube, 100mm, outside diameter 2.29mm, inside diameter 1.27mm, wall thickness 0.51mm, 99.95+%
Cd
GF03378652
Cadmium, rod, 100mm, diameter 5.0mm, as drawn, 99.99+%
Cd
GF50990597
Cadmium, foil, 150x150mm, thickness 0.25mm, as rolled, 99.95%
Cd
GF12224078
Cadmium, wire reel, 0.2m, diameter 0.25mm, hard, 99.99+%
Cd
GF79039648
Cadmium, rod, 50mm, diameter 10.0mm, 99.999%
Cd
GF37213893
Cadmium, wire reel, 0.2m, diameter 0.5mm, hard, 99.99+%
Cd
05-0075
Cadmium, ≥99.9%
Cd
GF10741889
Cadmium, foil, 100x100mm, thickness 3.0mm, as rolled, 99.9%
Cd
GF37649100
Cadmium, foil, 100x200mm, thickness 0.125mm, as rolled, 99.95%
Cd
GF38583287
Cadmium, foil, 100x300mm, thickness 0.125mm, as rolled, 99.95%
Cd
GF66037754
Cadmium, foil, 150x150mm, thickness 1.0mm, as rolled, 99.95%
Cd