EMAIL THIS PAGE TO A FRIEND
Environmental monitoring and assessment
Contamination of apple orchard soils and fruit trees with copper-based fungicides: sampling aspects.
PMID 25407992
Abstract
Accumulations of copper in orchard soils and fruit trees due to the application of Cu-based fungicides have become research hotspots. However, information about the sampling strategies, which can affect the accuracy of the following research results, is lacking. This study aimed to determine some sampling considerations when Cu accumulations in the soils and fruit trees of apple orchards are studied. The study was conducted in three apple orchards from different sites. Each orchard included two different histories of Cu-based fungicides usage, varying from 3 to 28 years. Soil samples were collected from different locations varying with the distances from tree trunk to the canopy drip line. Fruits and leaves from the middle heights of tree canopy at two locations (outer canopy and inner canopy) were collected. The variation in total soil Cu concentrations between orchards was much greater than the variation within orchards. Total soil Cu concentrations had a tendency to increase with the increasing history of Cu-based fungicides usage. Moreover, total soil Cu concentrations had the lowest values at the canopy drip line, while the highest values were found at the half distances between the trunk and the canopy drip line. Additionally, Cu concentrations of leaves and fruits from the outer parts of the canopy were significantly higher than from the inner parts. Depending on the findings of this study, not only the between-orchard variation but also the within-orchard variation should be taken into consideration when conducting future soil and tree samplings in apple orchards.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
BCR058
Continuous cast copper (O), BCR® certified Reference Material, rod
Cu
31284
Copper, electrolytic, Analytical Reagent, Reag. Ph. Eur., ≥99.9% (complexometric), wire
Cu
326445
Copper, bars (random sizes), ≥99.999% trace metals basis
Cu
254177
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Cu
326488
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Cu
266744
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Cu
349178
Copper, foil, thickness 0.25 mm, 99.98% trace metals basis
Cu
349208
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Cu
12816
Copper, foil, ≥99.8% (complexometric)
Cu
203122
Copper, powder, 99.999% trace metals basis
Cu
266086
Copper, powder, <425 μm, 99.5% trace metals basis
Cu
207780
Copper, powder, <75 μm, 99%
Cu
326453
Copper, powder (spheroidal), 10-25 μm, 98%
Cu
357456
Copper, powder (dendritic), <45 μm, 99.7% trace metals basis
Cu
365327
Copper, rod, diam. 11 mm, 99.9999% trace metals basis
Cu
223409
Copper, shot, −3-+14 mesh, 99%
Cu
520381
Copper, spherical shot, −20-+30 mesh, ≥99.5% trace metals basis
Cu
349224
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Cu
349216
Copper, wire, diam. 1.0 mm, ≥99.9%
Cu
326410
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Cu
326429
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Cu
61139
Copper, turnings, purum p.a., ≥99.0%
Cu
61141
Copper, purum p.a., ≥99.0%, powder
Cu
GF14058705
Copper, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.999%
Cu
GF71667366
Copper, wire reel, 1000m, diameter 0.04mm, annealed, 99.9%
Cu
GF21709650
Copper, wire reel, 1000m, diameter 0.04mm, hard, 99.9%
Cu
GF99677165
Copper, wire reel, 1000m, diameter 0.05mm, annealed, 99.9%
Cu
GF08494577
Copper, wire reel, 1000m, diameter 0.075mm, annealed, 99.9%
Cu
GF66965956
Copper, wire reel, 1000m, diameter 0.125mm, annealed, 99.9%
Cu
GF22319872
Copper, wire reel, 1000m, diameter 0.15mm, annealed, silver plated, 99.9%
Cu