EMAIL THIS PAGE TO A FRIEND
Soft matter
A peptide from human semenogelin I self-assembles into a pH-responsive hydrogel.
PMID 25408475
Abstract
The peptide GSFSIQYTYHV derived from human semenogelin I forms a transparent hydrogel through spontaneous self-assembly in water at neutral pH. Linear rheology measurements demonstrate that the gel shows a dominating elastic response over a large frequency interval. CD, fluorescence and FTIR spectroscopy and cryo-TEM studies imply long fibrillar aggregates of extended β-sheet. Dynamic light scattering data indicate that the fibril lengths are of the order of micrometers. Time-dependent thioflavin T fluorescence shows that fibril formation by GSFSIQYTYHV is a nucleated reaction. The peptide may serve as basis for development of smart biomaterials of low immunogenicity suitable for biomedical applications, including drug delivery and wound healing.