PloS one
In vivo notch signaling blockade induces abnormal spermatogenesis in the mouse.
PMID 25412258
Abstract
In a previous study we identified active Notch signaling in key cellular events occurring at adult spermatogenesis. In this study, we evaluated the function of Notch signaling in spermatogenesis through the effects of in vivo Notch blockade. Adult CD1 male mice were either submitted to a long term DAPT (?-secretase inhibitor) or vehicle treatment. Treatment duration was designed to attain one half the time (25 days) or the time (43 days) required to accomplish a complete cycle of spermatogenesis. Blockade of Notch signaling was depicted from decreased transcription of Notch effector genes. Notch signaling blockade disrupted the expression patterns of Notch components in the testis, induced male germ cell fate aberrations, and significantly increased germ cell apoptosis, mainly in the last stages of the spermatogenic cycle, and epididymis spermatozoa morphological defects. These effects were more pronounced following the 43 day than the 25 day DAPT treatment schedule. These results indicate a relevant regulatory role of Notch signaling in mammalian spermatogenesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA055704
Anti-CHST3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1306994
ANTI-CHST3(N-TERMINAL) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
WH0009469M1
Monoclonal Anti-CHST3 antibody produced in mouse, clone 1C4, purified immunoglobulin, buffered aqueous solution
SAB1401475
Monoclonal Anti-CHST3 antibody produced in mouse, clone 1D3, purified immunoglobulin, buffered aqueous solution
A8062 Monoclonal Anti-Goat/Sheep IgG–Alkaline Phosphatase antibody produced in mouse, clone GT-34, purified immunoglobulin, buffered aqueous glycerol solution
B3148 Monoclonal Anti-Goat/Sheep IgG–Biotin antibody produced in mouse, clone GT-34, purified immunoglobulin, buffered aqueous solution
F4891 Monoclonal Anti-Goat/Sheep IgG−FITC antibody produced in mouse, clone GT-34, purified immunoglobulin, buffered aqueous solution
A9452 Monoclonal Anti-Goat/Sheep IgG–Peroxidase antibody produced in mouse, clone GT-34, purified immunoglobulin, lyophilized powder
T1500
Testosterone, ≥98%
C19H28O2
T0100000
Testosterone, European Pharmacopoeia (EP) Reference Standard
C19H28O2
46923
Testosterone, VETRANAL, analytical standard
C19H28O2
Y0000342
Testosterone for impurity D identification, European Pharmacopoeia (EP) Reference Standard
C19H28O2
Y0000343
Testosterone for system suitability, European Pharmacopoeia (EP) Reference Standard
C19H28O2
T-037
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
C19H28O2