EMAIL THIS PAGE TO A FRIEND
Cell Death & Disease
Combination simvastatin and metformin induces G1-phase cell cycle arrest and Ripk1- and Ripk3-dependent necrosis in C4-2B osseous metastatic castration-resistant prostate cancer cells.
PMID 25412314