EMAIL THIS PAGE TO A FRIEND
Nature Communications
Bacteria slingshot more on soft surfaces.
PMID 25412641
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
282626
Trimethoxysilane, 95%
C3H10O3Si