EMAIL THIS PAGE TO A FRIEND
Echocardiography (Mount Kisco, N.Y.)
Effects of Insulin Resistance on Myocardial Blood Flow and Arterial Peripheral Circulation in Patients with Polycystic Ovary Syndrome.
PMID 25412756
Abstract
Polycystic ovary syndrome (PCOS) is associated with increased risk for cardiovascular disease. We sought to evaluate the effects of insulin resistance (IR) on myocardial microcirculation and peripheral artery function in patients with PCOS. We studied 55 women (28 with PCOS without IR, 18 with PCOS and IR and 11 normal controls) who underwent laboratorial analysis, high-resolution vascular ultrasound and real time myocardial contrast echocardiography (RTMCE). Intima-media thickness (IMT) and brachial artery flow-mediated dilation (FMD) were evaluated by vascular ultrasound. The replenishment velocity (β), plateau of acoustic intensity (A) and myocardial blood flow reserve (MBFR) were determined by quantitative dipyridamole stress RTMCE. β reserve in group PCOSxa0+xa0IR was lower than control (2.34xa0±xa00.55 vs. 3.60xa0±xa00.6; Pxa0