EMAIL THIS PAGE TO A FRIEND
Diabetes
Response to Comment on Sato et al. Improving type 2 diabetes through a distinct adrenergic signaling pathway involving mTORC2 that mediates glucose uptake in skeletal muscle. Diabetes 2014;63:4115-4129.
PMID 25414024