EMAIL THIS PAGE TO A FRIEND
Scientific reports
Heterogeneous red blood cell adhesion and deformability in sickle cell disease.
PMID 25417696
Abstract
We present a microfluidic approach that allows simultaneous interrogation of RBC properties in physiological flow conditions at a single cell level. With this method, we studied healthy hemoglobin A (HbA) and homozygous sickle hemoglobin (HbS) containing RBCs using whole blood samples from twelve subjects. We report that HbS-containing RBCs are heterogeneous in terms of adhesion and deformability in flow.