EMAIL THIS PAGE TO A FRIEND
Toxicology mechanisms and methods
Altered oxidative status and integrin expression in cyclosporine A-treated oral epithelial cells.
PMID 25418342
Abstract
Cyclosporine A (CsA) is an immunosuppressive agent administered to transplant patients. A well-known reported oral side effect of CsA consumption is gingival overgrowth (GO). Changes in the expression of integrins occurring in the gingiva following CsA treatment have been reported but these reports are mainly concerned with the connective tissue of the gingiva. In this study we targeted the alterations in the oral epithelium using KB cells, an oral epithelial cell line. Cultured oral epithelial cells were treated with increasing concentrations of CsA (0.1, 1 and 10 µg/mL) and the molecular changes involving antioxidant enzymes [glutathione peroxidase (GPx) and glutathione reductase (GR)] and the level of reactive oxygen species (ROS) were measured. Quantitative real-time PCR was used to assess the expression of selected integrins (α2, α5 and β1). At CsA concentration above 0.1 µg/mL GPx demonstrated an increase in activity while GR activity and the level of reduced glutathione were diminished (p < 0.05). α5 and β1 integrin were downregulated at all treatment concentrations of CsA while α2 integrin presented this effect at concentrations above 1 µg/mL (p < 0.05). The results suggest a possible role for oxidative stress and the altered expression of integrins in the pathology of CsA-induced gingival overgrowth.