EMAIL THIS PAGE TO A FRIEND
ACS chemical biology
Pseudoaeruginosins, nonribosomal peptides in Nodularia spumigena.
PMID 25419633
Abstract
Nodularia spumigena is a filamentous cyanobacterium that forms toxic blooms in brackish waters around the world through the production of the pentapeptide toxin nodularin. This cyanobacterium also produces large amounts of protease inhibitors belonging to the aeruginosin and spumigin families. Here we report the discovery of previously unknown protease inhibitors, pseudoaeruginosins NS1 (1) and NS2 (2), from 33 strains of N. spumigena isolated from the Baltic Sea. Pseudoaeruginosin NS1 (1) and NS2 (2) contain hexanoic acid, tyrosine, 4-methylproline, and argininal/argininol. The chemical structure of the two pseudoaeruginosins was verified by thorough comparison of the liquid chromatography-mass spectrometry (LC-MS) analyses of the extracts from the N. spumigena strains with synthetic peptides. The structures of the synthetic pseudoaeruginosins were confirmed using nuclear magnetic resonance spectroscopy. Surprisingly, the structure of pseudoaeruginosin NS1 (1) and NS2 (2) combines features of both aeruginosins and spumigins, suggesting that they have been produced through the joint action of both the spumigin and aeruginosin biosynthesis pathways. We screened with polymerase chain reaction and LC-MS 68 N. spumigena strains from the Baltic Sea and Australia. Pseudoaeruginosins were present in half of the Baltic Sea strains but were not found from the Australian strains. The production of pseudoaeruginosin seems to be coupled to the production of aeruginosins and 4-methylproline-containing spumigins. Pseudoaeruginosin NS1 was found to be as potent trypsin inhibitor as the most potent aeruginosins and spumigins with an IC50 of 0.19 ± 0.04 μM. This finding suggests that cooperation between the spumigin and aeruginosin biosynthetic pathways results in hybrid pseudoaeruginosin peptides.