EMAIL THIS PAGE TO A FRIEND
Alcoholism, Clinical and Experimental Research
Ethanol attenuation of long-term depression in the nucleus accumbens can be overcome by activation of TRPV1 receptors.
PMID 25421513