EMAIL THIS PAGE TO A FRIEND
Cancer chemotherapy and pharmacology
Phase I trial of S-1 every other day in combination with gemcitabine/cisplatin for inoperable biliary tract cancer.
PMID 25422155
Abstract
To date, gemcitabine-based or fluoropyrimidine-based regimens are recommended for unresectable advanced biliary tract cancer. Then, we conducted a phase I study of gemcitabine/cisplatin and S-1 that is an oral fluoropyrimidine. The aim of this study was to determine the dose-limiting toxicity (DLT), maximum-tolerated dose, and a recommended phase II dose of S-1. Response was assessed as a secondary endpoint. Patients who have been diagnosed with unresectable or postoperative recurrent biliary tract cancer received cisplatin (25 mg/m² i.v. for 120 min) followed by gemcitabine (1,000 mg/m² i.v. for 30 min) on days 1 and 8, and oral S-1 on alternate days; this regimen was repeated at 21-day intervals. A standard '3 + 3' phase I dose-escalation design was adopted. This study was registered with University hospital Medical Information Network (UMIN) Center in Japan, number UMIN000008415. Twelve patients were evaluable in this study. No patients developed DLTs. Recommended dose of S-1 was 80 (<1.25 m²), 100 (1.25 ≤ 1.5 m²), and 120 mg (1.5 m²≥) per day. One patient could achieve conversion to curative surgery. This phase I study was performed safely and demonstrated encouraging response.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T7205
Tegafur, ≥98% (HPLC), powder
C8H9FN2O3