EMAIL THIS PAGE TO A FRIEND
The Journal of cell biology
Dynein light intermediate chains maintain spindle bipolarity by functioning in centriole cohesion.
PMID 25422374
Abstract
Cytoplasmic dynein 1 (dynein) is a minus end-directed microtubule motor protein with many cellular functions, including during cell division. The role of the light intermediate chains (LICs; DYNC1LI1 and 2) within the complex is poorly understood. In this paper, we have used small interfering RNAs or morpholino oligonucleotides to deplete the LICs in human cell lines and Xenopus laevis early embryos to dissect the LICs' role in cell division. We show that although dynein lacking LICs drives microtubule gliding at normal rates, the LICs are required for the formation and maintenance of a bipolar spindle. Multipolar spindles with poles that contain single centrioles were formed in cells lacking LICs, indicating that they are needed for maintaining centrosome integrity. The formation of multipolar spindles via centrosome splitting after LIC depletion could be rescued by inhibiting Eg5. This suggests a novel role for the dynein complex, counteracted by Eg5, in the maintenance of centriole cohesion during mitosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32883
4-tert-Octylphenol monoethoxylate solution, 1 μg/mL in acetone, analytical standard
C16H26O2
32882
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
C16H26O2
A9251
Adenosine, ≥99%
C10H13N5O4
A4036
Adenosine, BioReagent, suitable for cell culture
C10H13N5O4
01890
Adenosine
C10H13N5O4
A0230200
Adenosine, European Pharmacopoeia (EP) Reference Standard
C10H13N5O4
PHR1138
Adenosine, Pharmaceutical Secondary Standard; Certified Reference Material
C10H13N5O4
SAB1401117
Anti-DYNC1LI2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
305197
Benzyl alcohol, anhydrous, 99.8%
C7H8O
ARK2163
Benzyl alcohol, PharmaGrade, USP/NF, Ph Eur, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
C7H8O
39971
Benzyl alcohol, certified reference material, TraceCERT®
C7H8O
08421
Benzyl alcohol, analytical standard
C7H8O
1061901
Benzyl alcohol, United States Pharmacopeia (USP) Reference Standard
C7H8O
13162
Benzyl alcohol, tested according to Ph.Eur.
C7H8O
PHR1019
Benzyl alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
C7H8O
W213705
Benzyl alcohol, ≥99%, FCC, FG
C7H8O
W213713
Benzyl alcohol, natural, ≥98%, FG
C7H8O
Y0000167
Benzyl alcohol, European Pharmacopoeia (EP) Reference Standard
C7H8O
C8273
Cytochalasin D, from Zygosporium mansonii, ≥98% (TLC and HPLC), powder
C30H37NO6
C2618
Cytochalasin D, Ready Made Solution, from Zygosporium mansonii, 5 mg/mL in DMSO, 0.2 μm filtered
C30H37NO6
94563
Dimethyl sulfoxide, analytical standard
C2H6OS
01934
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
C2H6OS
1211006
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
C2H6OS
D2050000
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
C2H6OS
T7402
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder
C47H51NO14
T1912
Paclitaxel, from Taxus yannanensis, powder
C47H51NO14
T7191
Paclitaxel, from semisynthetic (from Taxus sp.), ≥97%
C47H51NO14
Y0000698
Paclitaxel, European Pharmacopoeia (EP) Reference Standard
C47H51NO14
Y0000700
Paclitaxel natural for peak identification, European Pharmacopoeia (EP) Reference Standard
C47H51NO14
Y0000719
Paclitaxel semi-synthetic for peak identification, European Pharmacopoeia (EP) Reference Standard
C47H51NO14