EMAIL THIS PAGE TO A FRIEND
Iranian Journal of Basic Medical Sciences
The role of local renin-angiotensin system on high glucose-induced cell toxicity, apoptosis and reactive oxygen species production in PC12 cells.
PMID 25422756