EMAIL THIS PAGE TO A FRIEND
BMC Cancer
Calmodulin antagonists induce cell cycle arrest and apoptosis in vitro and inhibit tumor growth in vivo in human multiple myeloma.
PMID 25424011
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3281
N-(6-Aminohexyl)-5-chloro-1-naphthalenesulfonamide hydrochloride
C16H21ClN2O2S · HCl
SML0717
W-13, ≥98% (HPLC)
C14H17ClN2O2S · HCl