EMAIL THIS PAGE TO A FRIEND
Endocrine
Upregulation of podocyte-secreted angiopoietin-like-4 in diabetic nephropathy.
PMID 25424436
Abstract
Podocyte injury plays a key role in the development of diabetic nephropathy (DN). Understanding the changes in podocyte structure and function in diabetes mellitus may lead to novel diagnostic tools and treatment strategies for DN. Albuminuria, histological alterations, and podocyte injury were detected at different time points in streptozotocin (STZ)-induced diabetic rats. Increased urinary albumin-to-creatinine ratios (ACR) and podocyte injury were significantly observed 4xa0weeks post-STZ injection. We determined the glomerular expression and distribution of angiopoietin-like 4 (Angptl4) by immunofluorescence and real-time PCR. Glomerular Angptl4 expression was mostly colocalized with synaptopodin, a podocyte marker, with substantial additional overlap with the glomerular basement membrane (GBM). This finding indicated that Angptl4 might be primarily secreted by podocytes and moved toward the GBM. Moreover, we observed by Western blot analysis and ELISA that the urinary Angptl4 level was gradually upregulated in both STZ-induced rats and diabetic patients with microalbuminuria and macroalbuminuria. We further found that the increased glomerular Angptl4 expression was closely related to the urinary ACR level and podocyte injury. In addition, the urinary Angptl4 expression was closely associated with albuminuria in the rats and patients with DN. This study is the first to show that podocyte-secreted Angptl4 is upregulated in DN and can be detected in urine. Angptl4 might function as a podocyte injury marker and could be a potential and novel diagnostic and therapeutic biomarker for DN.