EMAIL THIS PAGE TO A FRIEND
Clinical and experimental pharmacology & physiology
β-asarone and levodopa co-administration protects against 6-hydroxydopamine-induced damage in parkinsonian rat mesencephalon by regulating autophagy: down-expression Beclin-1 and light chain 3B and up-expression P62.
PMID 25424835
Abstract
In this study, we investigated Beclin-1, light chain (LC)3B, and p62 expression in 6-hydroxydopamine (6-OHDA)-induced parkinsonian rats after β-asarone and levodopa (l-dopa) co-administration. Unilateral 6-OHDA injection into the medial forebrain bundle was used to create the models, except in sham-operated rats. Rats were divided into eight groups: sham-operated group; 6-OHDA model group; madopar group (75 mg/kg, per os (p.o.)); l-dopa group (60 mg/kg, p.o.); β-asarone group (15 mg/kg, p.o.); β-asarone + l-dopa co-administered group (15 mg/kg + 60 mg/kg, p.o.); 3-methyladenine group (500 nmol, intraperitoneal injection); and rapamycin group (1 mg/kg, intraperitoneal injection). Then, Beclin-1, LC3B, and p62 expression in the mesencephalon were detected. The mesencephalon was also observed by transmission electron microscope. The results showed that Beclin-1 and LC3B expression decreased and that p62 expression increased significantly in the madopar, l-dopa, β-asarone, and co-administered groups when compared with the 6-OHDA model. Beclin-1 and LC3B expression in the β-asarone and co-administered groups were less than in the madopar or l-dopa groups, whereas p62 expression in the β-asarone and co-administered groups was higher than in the madopar or l-dopa groups. In addition, a significant decrease in autophagosome was exhibited in the β-asarone and co-administered groups when compared with the 6-OHDA group. Our findings indicate that Beclin-1 and LC3B expression decreased, whereas p62 expression increased after co-administration treatment. In sum, all data suggest that the co-administration of β-asarone and l-dopa may contribute to the treatment of 6-OHDA-induced damage in rats by inhibiting autophagy activity.