EMAIL THIS PAGE TO A FRIEND
Nucleic Acids Research
Loss of loop adenines alters human telomere d[AG3(TTAG3)3] quadruplex folding.
PMID 25428355
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E9379
Stains-All, ~95%
C30H27BrN2S2
230960
Stains-All, Dye content 95 %
C30H27BrN2S2