EMAIL THIS PAGE TO A FRIEND
The Journal of Investigative Dermatology
Electrical stimulation enhances epidermal proliferation in human cutaneous wounds by modulating p53-SIVA1 interaction.
PMID 25431847