EMAIL THIS PAGE TO A FRIEND
Cancer research
Akt inhibition enhances expansion of potent tumor-specific lymphocytes with memory cell characteristics.
PMID 25432172
Abstract
Adoptive cell therapy (ACT) using autologous tumor-infiltrating lymphocytes (TIL) results in complete regression of advanced cancer in some patients, but the efficacy of this potentially curative therapy may be limited by poor persistence of TIL after adoptive transfer. Pharmacologic inhibition of the serine/threonine kinase Akt has recently been shown to promote immunologic memory in virus-specific murine models, but whether this approach enhances features of memory (e.g., long-term persistence) in TIL that are characteristically exhausted and senescent is not established. Here, we show that pharmacologic inhibition of Akt enables expansion of TIL with the transcriptional, metabolic, and functional properties characteristic of memory T cells. Consequently, Akt inhibition results in enhanced persistence of TIL after adoptive transfer into an immunodeficient animal model and augments antitumor immunity of CD8 T cells in a mouse model of cell-based immunotherapy. Pharmacologic inhibition of Akt represents a novel immunometabolomic approach to enhance the persistence of antitumor T cells and improve the efficacy of cell-based immunotherapy for metastatic cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45656
Rotenone, PESTANAL®, analytical standard
C23H22O6