EMAIL THIS PAGE TO A FRIEND
Journal of Pharmaceutical and Biomedical Analysis
Innovative sampling and extraction methods for the determination of nonsteroidal anti-inflammatory drugs in water.
PMID 25434009