EMAIL THIS PAGE TO A FRIEND
Microbial Pathogenesis
Differential gene expression profiling of Actinobacillus pleuropneumoniae during induction of primary alveolar macrophage apoptosis in piglets.
PMID 25435362