EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography A
Comparative evaluation of eight software programs for alignment of gas chromatography-mass spectrometry chromatograms in metabolomics experiments.
PMID 25435458