EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography A
Validation of a rapid method of analysis using ultrahigh-performance liquid chromatography - tandem mass spectrometry for nitrogen-rich adulterants in nutritional food ingredients.
PMID 25435465