EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences
A novel method for the simultaneous analysis of seven biothiols in rice (Oryza sativa L.) using hydrophilic interaction chromatography coupled with electrospray tandem mass spectrometry.
PMID 25436484