EMAIL THIS PAGE TO A FRIEND
Psychoneuroendocrinology
Maternal salivary cortisol levels during pregnancy are positively associated with overweight children.
PMID 25437119
Abstract
Animal and human studies suggest that programing of the hypothalamic-pituitary-adrenal (HPA) axis may be involved in the development of obesity, but human studies of biological indicators of HPA axis activity are lacking. We studied the association between levels of the stress hormone cortisol during pregnancy and overweight offspring during childhood into adolescence. Salivary samples from 655 Danish pregnant women with singleton pregnancies (1989-1991) were collected once in the morning and once in the evening in their second and third trimester. We followed the offspring from two to 16 years of age with at least one measurement of height and weight, and classified their body mass index into overweight and normal weight. The adjusted relative difference in median salivary cortisol (with 95% confidence interval (CI)) during pregnancy (the four samples), in second and third trimester (morning and evening samples) between overweight and normal weight offspring was estimated. Furthermore, the adjusted median ratio between morning and evening maternal salivary cortisol level was estimated for normal weight and overweight children. All the analyses were stratified into the equal age groups: 2-6, 7-11, and 12-16 years. We found non-significant higher maternal cortisol levels during pregnancy in offspring that were overweight at the age of 2-6, 7-11 and 12-16 years than in normal weight peers; adjusted relative difference in median salivary cortisol 11% (95% CI: -2; 25), 6% (95% CI: -7; 20), and 9% (95% CI: -4; 24), respectively. A statistically significantly higher level of maternal cortisol was found in the second trimester in 2-6-year-old and 12-16-year-old overweight offspring; relative difference 19% (95% CI: 3; 37), and 20% (95% CI: 3; 41), respectively. The median ratio between morning and evening maternal salivary cortisol level was similar for overweight and normal weight children; e.g. at age 2-6 years in third trimester 4.31 (95% CI: 4.05; 4.60)nmol/l and 4.28 (95% CI: 3.60; 5.09)nmol/l, respectively (P=0.93). Our findings suggest a relatively consistent association between pregnancy cortisol levels and overweight offspring, especially in the second trimester.