EMAIL THIS PAGE TO A FRIEND
Critical Care and Resuscitation : Journal of the Australasian Academy of Critical Care Medicine
A protocol for the 0.9% saline versus Plasma-Lyte 148 for intensive care fluid therapy (SPLIT) study.
PMID 25437221