EMAIL THIS PAGE TO A FRIEND
Laboratory Investigation; a Journal of Technical Methods and Pathology
Hemoglobin-associated oxidative stress in the pericardial compartment of postoperative cardiac surgery patients.
PMID 25437645