EMAIL THIS PAGE TO A FRIEND
Atherosclerosis
Circulating retinol binding protein 4 is associated with coronary lesion severity of patients with coronary artery disease.
PMID 25437889
Abstract
This study was to examine the association between plasma retinol binding protein 4 (RBP4) levels and the complexity of angiographic coronary lesion in patients with coronary artery disease (CAD). A cross-sectional and prospective study was carried out in Guangzhou Chinese population. 672 persons were evaluated by medical history, clinical examination, coronary angiography, and fasting plasma samples, and were followed prospectively for 3 years. We measured the plasma RBP4 levels in 447 women (201 with stable CAD and 246 with acute coronary syndrome [ACS]). Coronary lesions were classified as having a simple or complex appearance based on the visual estimation of the coronary angiograms. Median plasma RBP4 levels were significantly higher in stable CAD patients with complex coronary lesions (nxa0=xa084) than in those with simple lesions (nxa0=xa0117) (38.78[range 32.65-46.91] vs. 30.78 [range 24.48-36.08] μg/ml, Pxa0