EMAIL THIS PAGE TO A FRIEND
The Journal of surgical research
Orthotopic pancreatic tumors detected by optoacoustic tomography using Syndecan-1.
PMID 25439222
Abstract
Advances in small animal imaging have improved the detection and monitoring of cancer inxa0vivo; although with orthotopic models, precise localization of tumors remains a challenge. In this study, we evaluated multispectral optoacoustic tomography (MSOT) as an imaging modality to detect pancreatic adenocarcinoma in an orthotopic murine model. Inxa0vitro binding of Syndecan-1 probe to the human pancreatic cancer cell line S2VP10 was evaluated on flow cytometry. For inxa0vivo testing, S2VP10 cells were orthotopically implanted into the pancreas of severe combined immunodeficiency mice. At 7xa0d after implantation, the mice were intravenously injected with Syndecan-1 probe, and tumor uptake was evaluated with MSOT at multiple time points. Comparison was made with a free-dye control, indocyanine green (ICG). Probe uptake was verified exxa0vivo with fluorescent imaging. Syndecan-1 probe demonstrated partial binding to S2VP10 cells inxa0vitro. Inxa0vivo, Syndecan-1 probe preferentially accumulated in the pancreas tumor (480 MSOT a.u.) compared with off-target organs, including the liver (67 MSOT a.u.) and kidney (96 MSOT a.u.). Syndecan-1 probe accumulation peaked at 6xa0h (480 MSOT a.u.), whereas the ICG control dye failed to demonstrate similar retention within the tumor bed (0.0003 MSOT a.u.). At peak accumulation, signal intensity was 480 MSOT a.u., resulting in several times greater signal in the tumor bed than in the kidney or liver. Exxa0vivo fluorescent imaging comparing tumor signal with that within off-target organs confirmed the inxa0vivo results. MSOT demonstrates successful accumulation of Syndecan-1 probe within pancreatic tumors, and provides high-resolution images, which allow noninvasive, real-time comparison of signal within individual organs. Syndecan-1 probe preferentially accumulates within a pancreatic adenocarcinoma model, with minimal off-target effects.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
21980
Cardiogreen, for microscopy
C43H47N2NaO6S2