EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Evaluation and comparison of the kinetic performance of ultra-high performance liquid chromatography and high-performance liquid chromatography columns in hydrophilic interaction and reversed-phase liquid chromatography conditions.
PMID 25441074
Abstract
An intrinsic performance comparison is made of the reduction in analysis time that can be obtained when switching from HPLC to UHPLC column formats in HILIC and reversed-phase conditions. A detailed overview of the packing characteristics of both stationary phase types is given first. It is demonstrated that HILIC columns demonstrate higher external porosity values than their reversed-phase counterparts resulting in lower flow resistance values. Column total porosity values determined from the elution time of a small marker molecule are shown to depend strongly on the composition of the mobile phase. To omit errors that might arise from an over- or underestimation of the column void time, all plate height and kinetic plot data are therefore expressed as a function of the interstitial velocity. Although only a limited number of columns are evaluated in this study, it is shown that the column efficiency of the HILIC columns is lower than that of their reversed-phase counterparts, at least for the compounds evaluated here. Despite this lower efficiency, the kinetic performance of both stationary phase types is similar, due to the much lower viscosity of the mobile phases typically used in HILIC conditions. Finally, it is demonstrated that a similar, yet slightly larger reduction in analysis time can be obtained when switching from HPLC column formats to UHPLC formats in HILIC compared to reversed-phase conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31573
5α-Androstan-17β-ol-3-one, VETRANAL, analytical standard
C19H30O2
215376
Acenaphthene, 99%
C12H10
05426
Acenaphthene, certified reference material, TraceCERT®
C12H10
48500-U
Acenaphthene, analytical standard
C12H10
A8626
Adenine, ≥99%
C5H5N5
A2786
Adenine, suitable for cell culture, BioReagent
C5H5N5
A5665
Adenine, BioReagent, plant cell culture tested, ≥99%
C5H5N5
A0230000
Adenine, European Pharmacopoeia (EP) Reference Standard
C5H5N5
PHR1383
Adenine, Pharmaceutical Secondary Standard; Certified Reference Material
C5H5N5
A9251
Adenosine, ≥99%
C10H13N5O4
A4036
Adenosine, suitable for cell culture, BioReagent
C10H13N5O4
01890
Adenosine
C10H13N5O4
A0230200
Adenosine, European Pharmacopoeia (EP) Reference Standard
C10H13N5O4
PHR1138
Adenosine, Pharmaceutical Secondary Standard; Certified Reference Material
C10H13N5O4
A1542
Ammonium acetate, for molecular biology, ≥98%
C2H7NO2
A7330
Ammonium acetate, BioXtra, ≥98%
C2H7NO2
09689
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
C2H7NO2
A7262
Ammonium acetate, reagent grade, ≥98%
C2H7NO2
73594
Ammonium acetate, for mass spectrometry, LiChropur, eluent additive for LC-MS
C2H7NO2
A2706
Ammonium acetate solution, for molecular biology, 7.5 M
C2H7NO2
09691
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
C2H7NO2
90577
Bifido Selective Supplement B, for microbiology
C2H4O2
C3506
Cytosine, ≥99%
C4H5N3O
PHR1350
Cytosine, Pharmaceutical Secondary Standard; Certified Reference Material
C4H5N3O
154792
Dichloromethane, ACS spectrophotometric grade, ≥99.5%, contains 50-150 ppm amylene as stabilizer
CH2Cl2
66742
Dichloromethane, Selectophore, ≥99.5%
CH2Cl2
02575
Dichloromethane, analytical standard
CH2Cl2
48538
Dichloromethane, analytical standard, stabilized
CH2Cl2
48600
Dichloromethane solution, certified reference material, 200 μg/mL in methanol
CH2Cl2
40042
Dichloromethane solution, certified reference material, 5000 μg/mL in methanol
CH2Cl2