EMAIL THIS PAGE TO A FRIEND
Analytica Chimica Acta
Ultra-sensitive determination of cyanide in surface water by gas chromatography-tandem mass spectrometry after derivatization with 2-(dimethylamino)ethanethiol.
PMID 25441894