EMAIL THIS PAGE TO A FRIEND
Letters in Applied Microbiology
Aeromonas piscicola AH-3 expresses an extracellular collagenase with cytotoxic properties.
PMID 25443157
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F5135
N-[3-(2-Furyl)acryloyl]-Leu-Gly-Pro-Ala
C23H32N4O7