EMAIL THIS PAGE TO A FRIEND
Cancer Letters
Metastasis-associated in colon cancer-1 upregulates vascular endothelial growth factor-C/D to promote lymphangiogenesis in human gastric cancer.
PMID 25444928