EMAIL THIS PAGE TO A FRIEND
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V
A novel oxido-viscosifying Hyaluronic Acid-antioxidant conjugate for osteoarthritis therapy: biocompatibility assessments.
PMID 25445302
Abstract
To overcome the problem of fast degradation of Hyaluronic Acid (HA) in the treatment of osteoarthritis (OA), HA was protected against the oxidative stress generated by the pathology. Antioxidant conjugated HAs were synthesized and tested in vitro for their resistance in an oxidative environment mimicking OA. HA-4-aminoresorcinol (HA-4AR) displayed the interesting property of increasing in viscosity under oxidative conditions because of crosslinking induced by electron transfer. The novel HA polymer conjugate was shown to be biocompatible in vitro on fibroblast-like synoviocytes extracted from an arthritic patient. This HA conjugate was also assessed in vivo by intra-articular injection in healthy rabbits and was found to be comparable to the native polymer in terms of biocompatibility. This study suggests that HA-4AR is a promising candidate for a next generation viscosupplementation formulation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
02562
1,2-Dichloroethane, analytical standard
C2H4Cl2