EMAIL THIS PAGE TO A FRIEND
Neuroscience Letters
The coexistence of VGluT2 and neurotensin or leu-enkephalin in the medullary dorsal horn: a confocal and electron microscopic immunohistochemical study in the rat.
PMID 25445367