EMAIL THIS PAGE TO A FRIEND
The Journal of antibiotics
Lanthiopeptin, a new peptide antibiotic. Production, isolation and properties of lanthiopeptin.
PMID 2544544
Abstract
A strain of Streptoverticillium cinnamoneum produced a peptide antibiotic named lanthiopeptin, which contained four unusual amino acids, erythro-beta-hydroxyaspartic acid, mesolanthionine, threo-beta-methyllanthionine and lysinoalanine. Lanthiopeptin showed antiviral activity against herpes simplex virus type 1 KOS strain infection in Vero cells by cytopathic effect reduction assay. The structure of lanthiopeptin is similar to that of ancovenin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C5241 Cinnamycin, from Streptomyces cinnamoneus, ≥95% (HPLC)
C89H125N25O25S3