EMAIL THIS PAGE TO A FRIEND
Progress in Neuro-Psychopharmacology & Biological Psychiatry
Blockade of spinal glutamate recycling produces paradoxical antinociception in rats with orofacial inflammatory pain.
PMID 25445477