EMAIL THIS PAGE TO A FRIEND
International journal of pharmaceutics
Protein release from electrospun nonwovens: improving the release characteristics through rational combination of polyester blend matrices with polidocanol.
PMID 25445519
Abstract
Nonwoven scaffolds consisting of poly-ε-caprolactone (PCL), poly(lactic-co-glycolic acid) (PLGA) and polidocanol (PD), and loaded with lysozyme crystals were prepared by electrospinning. The composition of the matrix was varied and the effect of PD content in binary mixtures, and of PD and PLGA content in ternary mixtures regarding processability, fiber morphology, water sorption, swelling and drug release was investigated. Binary PCL/PD blend nonwovens showed a PD-dependent increase in swelling of up to 30% and of lysozyme burst release of up to 45% associated with changes of the fiber morphology. Furthermore, addition of free PD to the release medium resulted in a significant increase of lysozyme burst release from pure PCL nonwovens from approximately 2-35%. Using ternary PCL/PD/PLGA blends, matrix degradation could be significantly improved over PCL/PD blends, resulting in a biphasic release of lysozyme with constant release over 9 weeks, followed by constant release with a reduced rate over additional 4 weeks. Based on these results, protein release from PCL scaffolds is improved by blending with PD due to improved lysozyme desorption from the polymer surface and PD-dependent matrix swelling.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P9641
Nonaethylene glycol monododecyl ether, nonionic surfactant
C30H62O10