EMAIL THIS PAGE TO A FRIEND
Toxicology Letters
Aryl hydrocarbon receptor SNP -130 C/T associates with dioxins susceptibility through regulating its receptor activity and downstream effectors including interleukin 24.
PMID 25445724